4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile
Supplier: Apollo Scientific
Warning
PC500654-1G PC500654-250MG
APOSPC500654-1GEA 905 EUR
APOSPC500654-1G APOSPC500654-250MG
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile
4-[1,1,2-Trifluoro-2-(trifluoromethoxy)ethoxy]benzonitrile
Formula: C₁₀H₅F₆NO₂
MW: 285.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103422
CAS Number: 1845713-67-8
UN: 3276
ADR: 6.1 III