Methyl 5-bromo-2-(trifluoromethyl)benzoate
Supplier: Apollo Scientific
PC500761-1G PC500761-250MG PC500761-5G
APOSPC500761-1GEA 145 EUR
APOSPC500761-1G APOSPC500761-250MG APOSPC500761-5G
Methyl 5-bromo-2-(trifluoromethyl)benzoate
Methyl 5-bromo-2-(trifluoromethyl)benzoate
Formula: C₉H₆BrF₃O₂
MW: 283.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14697999
CAS Number: 842136-32-7