2-(2,2-Difluoroethoxy)benzonitrile
Supplier: Apollo Scientific
Warning
PC500919-1G PC500919-5G
APOSPC500919-1GEA 197 EUR
APOSPC500919-1G APOSPC500919-5G
2-(2,2-Difluoroethoxy)benzonitrile
2-(2,2-Difluoroethoxy)benzonitrile
Formula: C₉H₇F₂NO
MW: 183.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699093
CAS Number: 1160246-19-4