5-(Difluoromethoxy)-2-fluorobenzaldehyde, tech
Supplier: Apollo Scientific
Warning
PC502194-100MG PC502194-250MG PC502194-1G
APOSPC502194-100MGEA 203 EUR
APOSPC502194-100MG APOSPC502194-250MG APOSPC502194-1G
5-(Difluoromethoxy)-2-fluorobenzaldehyde, tech
5-(Difluoromethoxy)-2-fluorobenzaldehyde
Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847161
CAS Number: 1214364-36-9