1-Bromo-4-(pentafluoroethoxy)benzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-(perfluoroethoxy)benzene
PC50221-1G PC50221-25G PC50221-5G
APOSPC50221-1GEA 36.4 EUR
APOSPC50221-1G APOSPC50221-25G APOSPC50221-5G
1-Bromo-4-(pentafluoroethoxy)benzene 99%
1-Bromo-4-(pentafluoroethoxy)benzene
Formula: C₈H₄BrF₅O
MW: 291.013 g/mol
Boiling Pt: 91…91 °C (50 mm Hg)
Density: 1.654 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040941
CAS Number: 56425-85-5