2-Bromo-6-(trifluoromethoxy)benzaldehyde
Supplier: Apollo Scientific
PC502684-1G PC502684-5G PC502684-10G
APOSPC502684-1GEA 60.9 EUR
APOSPC502684-1G APOSPC502684-5G APOSPC502684-10G
2-Bromo-6-(trifluoromethoxy)benzaldehyde
2-Bromo-6-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄BrF₃O₂
MW: 269.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856022
CAS Number: 1114809-17-4