2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Supplier: Apollo Scientific
Danger
PC502996-1G PC502996-25G PC502996-5G PC502996-100G
APOSPC502996-1GEA 87 EUR
APOSPC502996-1G APOSPC502996-25G APOSPC502996-5G APOSPC502996-100G
2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
2-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Formula: C₈H₃BrF₃NOS
MW: 298.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800656
CAS Number: 948294-38-0
UN: 2922
ADR: 8+6.1 III