2-Fluoro-3-(trifluoromethyl)toluene 98%
Supplier: Apollo Scientific
PC50415-10G PC50415-1G PC50415-5G
APOSPC50415-10GEA 763 EUR
APOSPC50415-10G APOSPC50415-1G APOSPC50415-5G
2-Fluoro-3-(trifluoromethyl)toluene 98%
2-Fluoro-3-(trifluoromethyl)toluene
Formula: C₈H₆F₄
MW: 178.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185663
CAS Number: 1214331-63-1