3,4-Dichloro-5-(trifluoromethoxy)bromobenzene 98%
Supplier: Apollo Scientific
Synonyms: 5-Bromo-1,2-dichloro-3-(trifluoromethoxy)benzene
PC50416-10G PC50416-1G PC50416-5G
APOSPC50416-10GEA 574 EUR
APOSPC50416-10G APOSPC50416-1G APOSPC50416-5G
3,4-Dichloro-5-(trifluoromethoxy)bromobenzene 98%
3,4-Dichloro-5-(trifluoromethoxy)bromobenzene
Formula: C₇H₂BrCl₂F₃O
MW: 309.895 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32708470
CAS Number: 2168733-41-1