2-Bromo-5-iodobenzotrifluoride
Supplier: Apollo Scientific
Warning
PC51000-1G PC51000-25G PC51000-5G
APOSPC51000-1GEA 21.9 EUR
APOSPC51000-1G APOSPC51000-25G APOSPC51000-5G
2-Bromo-5-iodobenzotrifluoride
2-Bromo-5-iodobenzotrifluoride
Formula: C₇H₃BrF₃I
Melting Pt: 78…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11855999
CAS Number: 364-11-4