2-(4-Bromophenyl)-2,2-difluoroacetic acid
Supplier: Apollo Scientific
Danger
PC510336-1G PC510336-250MG PC510336-5G
APOSPC510336-1GEA 219 EUR
APOSPC510336-1G APOSPC510336-250MG APOSPC510336-5G
2-(4-Bromophenyl)-2,2-difluoroacetic acid
2-(4-Bromophenyl)-2,2-difluoroacetic acid
Formula: C₈H₅BrF₂O₂
MW: 251.025 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD11167796
CAS Number: 913574-93-3
UN: 3261
ADR: 8 III