7-Bromo-5-(pentafluorothio)-1,3-benzoxazole 95%
Supplier: Apollo Scientific
Warning
Synonyms: 7-Bromo-5-(pentafluorosulphanyl)benzo[d]oxazole, (7-Bromo-1,3-benzoxazol-5-yl)sulphur pentafluoride
PC51189-1G PC51189-250MG
APOSPC51189-1GEA 1040 EUR
APOSPC51189-1G APOSPC51189-250MG
7-Bromo-5-(pentafluorothio)-1,3-benzoxazole 95%
7-Bromo-5-(pentafluorothio)-1,3-benzoxazole
Formula: C₇H₃BrF₅NOS
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD24369705
CAS Number: 1432075-78-9