1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-boronic acid, pinacol ester
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)-1H-pyrazole
PC512023-100MG PC512023-50MG
APOSPC512023-100MGEA 464 EUR
APOSPC512023-100MG APOSPC512023-50MG
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-boronic acid, pinacol ester
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-3-boronic acid, pinacol ester
Formula: C₁₁H₁₆BF₃N₂O₂
MW: 276.063 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28368565
CAS Number: 1396123-92-4