Methyl 2,3-difluoro-4-(trifluoromethoxy)benzoate 95%
Supplier: Apollo Scientific
Warning
PC51628-1G PC51628-250MG PC51628-5G
APOSPC51628-1GEA 58 EUR
APOSPC51628-1G APOSPC51628-250MG APOSPC51628-5G
Methyl 2,3-difluoro-4-(trifluoromethoxy)benzoate 95%
Methyl 2,3-difluoro-4-(trifluoromethoxy)benzoate
Formula: C₉H₅F₅O₃
MW: 256.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32066738
CAS Number: 2366994-13-8