Methyltrifluorosilane 98%

Supplier: Apollo Scientific
Warning

PC5186-10G PC5186-50G
APOSPC5186-10GEA 55.1 EUR
APOSPC5186-10G APOSPC5186-50G
Methyltrifluorosilane 98%
Methyltrifluorosilane
Formula: CH₃F₃Si
Boiling Pt: –30 °C
Melting Pt: –73 °C
Storage Temperature: Ambient
MDL Number: MFCD00040332
CAS Number: 373-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR