6-Fluoro-2-(trifluoromethyl)-1H-indole
Supplier: Apollo Scientific
PC52227-1G PC52227-250MG
APOSPC52227-1GEA 654 EUR
APOSPC52227-1G APOSPC52227-250MG
6-Fluoro-2-(trifluoromethyl)-1H-indole
6-Fluoro-2-(trifluoromethyl)-1H-indole
Formula: C₉H₅F₄N
MW: 203.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264122
CAS Number: 932014-36-3