2,4-Difluoro-5-(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-2,4-difluoro-5-(trifluoromethyl)benzene
PC53038-1G PC53038-250MG
APOSPC53038-1GEA 754 EUR
APOSPC53038-1G APOSPC53038-250MG
2,4-Difluoro-5-(trifluoromethyl)benzyl bromide
2,4-Difluoro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₄BrF₅
MW: 275.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28897198
CAS Number: 1934665-18-5
UN: 3265
ADR: 8 III