6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
PC53332-1G PC53332-250MG
APOSPC53332-1GEA 360 EUR
APOSPC53332-1G APOSPC53332-250MG
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine
Formula: C₇H₃BrF₃N₃
MW: 266.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111700
CAS Number: 1166819-53-9