(2S)-2-amino-3-(4-bromo-2-fluorophenyl)propanoic acid
Supplier: Apollo Scientific
Warning
PC53369-1G PC53369-5G
APOSPC53369-1GEA 161 EUR
APOSPC53369-1G APOSPC53369-5G
(2S)-2-amino-3-(4-bromo-2-fluorophenyl)propanoic acid
(2S)-2-amino-3-(4-bromo-2-fluorophenyl)propanoic acid
Formula: C₉H₉BrFNO₂
MW: 262.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14561453
CAS Number: 1213206-88-2