2-Methyl-2-[4-(trifluoromethoxy)phenoxy]acetic hydrazide 97%
Supplier: Apollo Scientific
Warning
PC5341-1G PC5341-250MG
APOSPC5341-1GEA 173 EUR
APOSPC5341-1G APOSPC5341-250MG
2-Methyl-2-[4-(trifluoromethoxy)phenoxy]acetic hydrazide 97%
2-Methyl-2-[4-(trifluoromethoxy)phenoxy]acetic hydrazide
Formula: C₁₀H₁₁F₃N₂O₃
MW: 264.201 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00052356
CAS Number: 175204-37-2