(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 97%
Supplier: Apollo Scientific
Warning
PC535001-100MG PC535001-250MG
APOSPC535001-100MGEA 92.8 EUR
APOSPC535001-100MG APOSPC535001-250MG
(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 97%
(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride
Formula: C₈H₉BrFN·ClH
MW: 254.527 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22988854
CAS Number: 1624261-91-1