(R)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 95+%
Supplier: Apollo Scientific
Warning
PC535002-100MG PC535002-250MG
APOSPC535002-100MGEA 87 EUR
APOSPC535002-100MG APOSPC535002-250MG
(R)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 95+%
(R)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride
Formula: C₈H₉BrFN·ClH
MW: 254.527 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22988853
CAS Number: 1624262-55-0