5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%
Supplier: Apollo Scientific
Warning
PC535088-100MG PC535088-250MG
APOSPC535088-100MGEA 21.9 EUR
APOSPC535088-100MG APOSPC535088-250MG
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one
Formula: C₉H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD18208183
CAS Number: 1260013-11-3