2-Bromo-3-hydroxy-6-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-6-(trifluoromethyl)pyridin-3-ol
PC53551-1G PC53551-250MG
APOSPC53551-1GEA 676 EUR
APOSPC53551-1G APOSPC53551-250MG
2-Bromo-3-hydroxy-6-(trifluoromethyl)pyridine
2-Bromo-3-hydroxy-6-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃NO
MW: 241.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15478054
CAS Number: 1211526-50-9