5-Fluoro-6-(trifluoromethyl)nicotinonitrile 95+%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Fluoro-6-(trifluoromethyl)pyridine-3-nitrile
PC56140-1G PC56140-250MG
APOSPC56140-1GEA 290 EUR
APOSPC56140-1G APOSPC56140-250MG
5-Fluoro-6-(trifluoromethyl)nicotinonitrile 95+%
5-Fluoro-6-(trifluoromethyl)nicotinonitrile
Formula: C₇H₂F₄N₂
MW: 190.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28770505
CAS Number: 1807303-10-1
UN: 3276
ADR: 6.1 III