1,1,1,2,2,3,3,4,4-Nonafluoro-6-(propoxy)hexane 99%
Supplier: Apollo Scientific
Warning
PC56214-5G PC56214-25G PC56214-100G
APOSPC56214-5GEA 115 EUR
APOSPC56214-5G APOSPC56214-25G APOSPC56214-100G
1,1,1,2,2,3,3,4,4-Nonafluoro-6-(propoxy)hexane 99%
1,1,1,2,2,3,3,4,4-Nonafluoro-6-(propoxy)hexane
Formula: C₉H₁₁F₉O
Boiling Pt: 142 °C
Melting Pt: –74 °C
Density: 1.3282 g/cm³ (25 °C)
Flash Pt: 41.5 °C
Storage Temperature: Ambient
MDL Number: MFCD23134588
CAS Number: 1193009-98-1
UN: 3271
ADR: 3,III