2,2-Difluoro-1-(4-fluorophenyl)ethanone 95%
Supplier: Apollo Scientific
Danger
PC56745-1G PC56745-250MG
APOSPC56745-1GEA 509 EUR
APOSPC56745-1G APOSPC56745-250MG
2,2-Difluoro-1-(4-fluorophenyl)ethanone 95%
2,2-Difluoro-1-(4-fluorophenyl)ethanone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618056
CAS Number: 50562-06-6