2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
Supplier: Apollo Scientific
Warning
PC56748-1G PC56748-250MG
APOSPC56748-1GEA 654 EUR
APOSPC56748-1G APOSPC56748-250MG
2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
2-Chloro-2,2-difluoro-1-(4-fluorophenyl)ethanone
Formula: C₈H₄ClF₃O
MW: 208.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921049
CAS Number: 1321028-35-6