2-Bromo-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
Supplier: Apollo Scientific
Warning
PC56749-1G PC56749-250MG
APOSPC56749-1GEA 654 EUR
APOSPC56749-1G APOSPC56749-250MG
2-Bromo-2,2-difluoro-1-(4-fluorophenyl)ethanone 95%
2-Bromo-2,2-difluoro-1-(4-fluorophenyl)ethanone
Formula: C₈H₄BrF₃O
MW: 253.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28408579
CAS Number: 1802650-67-4