7-(Trifluoromethoxy)-1H-indazole
Supplier: Apollo Scientific
Warning
PC56889-1G PC56889-250MG
APOSPC56889-1GEA 864 EUR
APOSPC56889-1G APOSPC56889-250MG
7-(Trifluoromethoxy)-1H-indazole
7-(Trifluoromethoxy)-1H-indazole
Formula: C₈H₅F₃N₂O
MW: 202.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31664915
CAS Number: 2244087-63-4