2-Fluoro-6-iodobenzyl alcohol
Supplier: Apollo Scientific
Warning
Synonyms: (2-Fluoro-6-iodophenyl)methanol
PC57046-1G PC57046-5G
APOSPC57046-1GEA 24.8 EUR
APOSPC57046-1G APOSPC57046-5G
2-Fluoro-6-iodobenzyl alcohol
2-Fluoro-6-iodobenzyl alcohol
Formula: C₇H₆FIO
MW: 252.025 g/mol
Melting Pt: 54…55 °C
Storage Temperature: Ambient
MDL Number: MFCD09910144
CAS Number: 911825-94-0