4-(tert-Butoxy)benzotrifluoride 97%
Supplier: Apollo Scientific
Synonyms: 1-(tert-Butoxy)-4-(trifluoromethyl)benzene, tert-Butyl 4-(trifluoromethyl)phenyl ether
PC5876-1G PC5876-25G PC5876-5G
APOSPC5876-1GEA 21.9 EUR
APOSPC5876-1G APOSPC5876-25G APOSPC5876-5G
4-(tert-Butoxy)benzotrifluoride 97%
4-(tert-Butoxy)benzotrifluoride
Formula: C₁₁H₁₃F₃O
MW: 218.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09763645
CAS Number: 16222-44-9