2-Phenoxy-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
PC6053-1G PC6053-5G
APOSPC6053-1GEA 72.5 EUR
APOSPC6053-1G APOSPC6053-5G
2-Phenoxy-5-(trifluoromethyl)pyridine
2-Phenoxy-5-(trifluoromethyl)pyridine
Formula: C₁₂H₈F₃NO
MW: 239.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07692289
CAS Number: 105626-77-5