2-Fluoro-3-(trifluoromethoxy)benzaldehyde
Supplier: Apollo Scientific
Warning
PC6309-1G PC6309-5G
APOSPC6309-1GEA 92.8 EUR
APOSPC6309-1G APOSPC6309-5G
2-Fluoro-3-(trifluoromethoxy)benzaldehyde
2-Fluoro-3-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026472
CAS Number: 1159512-58-9