2-Fluoro-3-(trifluoromethoxy)phenylacetic acid
Supplier: Apollo Scientific
Warning
PC6352-1G PC6352-5G
APOSPC6352-1GEA 279 EUR
APOSPC6352-1G APOSPC6352-5G
2-Fluoro-3-(trifluoromethoxy)phenylacetic acid
2-Fluoro-3-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD12026476
CAS Number: 1159512-61-4