Perfluoropolyether dicarboxylic acid

Supplier: Apollo Scientific
Danger

PC6422-1G PC6422-5G
APOSPC6422-1GEA 0 EUR
APOSPC6422-1G APOSPC6422-5G
Perfluoropolyether dicarboxylic acid
Perfluoropolyether dicarboxylic acid
Storage Temperature: Ambient
MDL Number: MFCD03100837
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR