O-(4-Bromo-2-fluorophenyl) dimethylthiocarbamate 98%

Supplier: Apollo Scientific

PC7279-1G PC7279-5G
APOSPC7279-1GEA 118 EUR
APOSPC7279-1G APOSPC7279-5G
O-(4-Bromo-2-fluorophenyl) dimethylthiocarbamate 98%
O-(4-Bromo-2-fluorophenyl) dimethylthiocarbamate
Formula: C₉H₉BrFNOS
Storage Temperature: Ambient
MDL Number: MFCD09258772
CAS Number: 190648-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR