2,5-Bis(trifluoromethyl)-7-chloro-1H-benzimidazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Bis(trifluoromethyl)-4-chloro-1H-benzimidazole
PC7695-1G PC7695-5G
APOSPC7695-1GEA 200 EUR
APOSPC7695-1G APOSPC7695-5G
2,5-Bis(trifluoromethyl)-7-chloro-1H-benzimidazole 97%
2,5-Bis(trifluoromethyl)-7-chloro-1H-benzimidazole
Formula: C₉H₃ClF₆N₂
MW: 288.577 g/mol
Melting Pt: 154…156 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD12547949
CAS Number: 1209789-75-2