(1H,1H,2H,2H-Perfluorotetradecyl)tris(ethoxy)silane
Supplier: Apollo Scientific
Warning
PC7904-25G PC7904-5G
APOSPC7904-25GEA 1540 EUR
APOSPC7904-25G APOSPC7904-5G
(1H,1H,2H,2H-Perfluorotetradecyl)tris(ethoxy)silane
(1H,1H,2H,2H-Perfluorotetradecyl)tris(ethoxy)silane
Formula: C₂₀H₁₉F₂₅O₃Si
Boiling Pt: 127…128 °C (0.2 mm Hg)
Melting Pt: 46…47 °C
Storage Temperature: Ambient
MDL Number: MFCD07368765
CAS Number: 885275-56-9