1-(2-Amino-4-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane hydrochloride
Supplier: Apollo Scientific
PC8134-1G PC8134-500MG
APOSPC8134-1GEA 799 EUR
APOSPC8134-1G APOSPC8134-500MG
1-(2-Amino-4-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane hydrochloride
1-(2-Amino-4-chlorophenylamino)-2-[3-chloro-5-(trifluoromethyl)pyridin-2-ylamino]ethane hydrochloride
Formula: C₁₄H₁₃Cl₂F₃N₄·ClH
MW: 401.642 g/mol
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00793025
CAS Number: 321385-54-0