5-(Bromomethyl)-2-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC8704-250MG PC8704-1G PC8704-5G
APOSPC8704-250MGEA 50.9 EUR
APOSPC8704-250MG APOSPC8704-1G APOSPC8704-5G
5-(Bromomethyl)-2-(trifluoromethyl)pyridine
5-(Bromomethyl)-2-(trifluoromethyl)pyridine
Formula: C₇H₅BrF₃N
MW: 240.021 g/mol
Melting Pt: 37…38 °C
Storage Temperature: Ambient
MDL Number: MFCD10697684
CAS Number: 108274-33-5