1,1,1,4,4,4-Hexafluorobutan-2-one
Supplier: Apollo Scientific
Warning
PC8839-1G PC8839-5G
APOSPC8839-1GEA 151 EUR
APOSPC8839-1G APOSPC8839-5G
1,1,1,4,4,4-Hexafluorobutan-2-one
1,1,1,4,4,4-Hexafluorobutan-2-one
Formula: C₄H₂F₆O
Boiling Pt: 54.2 °C (747 mm Hg)
Melting Pt: –56 °C
Density: 1.5589 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08460488
CAS Number: 400-49-7