1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid ≥98%
Supplier: Apollo Scientific
PC901560-1G PC901560-250MG PC901560-5G
APOSPC901560-1GEA 192 EUR
APOSPC901560-1G APOSPC901560-250MG APOSPC901560-5G
1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid ≥98%
1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD12405622
CAS Number: 872422-15-6