N-(3,5-difluorophenyl)formamide 95%
Supplier: Apollo Scientific
PC901747-10G PC901747-1G PC901747-5G
APOSPC901747-10GEA 957 EUR
APOSPC901747-10G APOSPC901747-1G APOSPC901747-5G
N-(3,5-difluorophenyl)formamide 95%
N-(3,5-difluorophenyl)formamide
Formula: C₇H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD00034516
CAS Number: 198077-69-9