1-(3-Fluorophenyl)-2,5-dimethylpyrrole 97%
Supplier: Apollo Scientific
Synonyms: 1-(3-Fluorophenyl)-2,5-dimethyl-1H-pyrrole
PC902044-1G PC902044-5G
APOSPC902044-1GEA 140 EUR
APOSPC902044-1G APOSPC902044-5G
1-(3-Fluorophenyl)-2,5-dimethylpyrrole 97%
1-(3-Fluorophenyl)-2,5-dimethylpyrrole
Formula: C₁₂H₁₂FN
MW: 189.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07429171
CAS Number: 146135-21-9