N-Cyclopropyl-5-fluoro-2-nitroaniline 97%
Supplier: Apollo Scientific
PC902322-10G PC902322-1G PC902322-5G
APOSPC902322-10GEA 502 EUR
APOSPC902322-10G APOSPC902322-1G APOSPC902322-5G
N-Cyclopropyl-5-fluoro-2-nitroaniline 97%
N-Cyclopropyl-5-fluoro-2-nitroaniline
Formula: C₉H₉FN₂O₂
MW: 196.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16744208
CAS Number: 1248276-34-7