2,3-Difluoro-N-methyl-6-nitroaniline 97%
Supplier: Apollo Scientific
PC902355-100G PC902355-25G PC902355-5G
APOSPC902355-100GEA 632 EUR
APOSPC902355-100G APOSPC902355-25G APOSPC902355-5G
2,3-Difluoro-N-methyl-6-nitroaniline 97%
2,3-Difluoro-N-methyl-6-nitroaniline
Formula: C₇H₆F₂N₂O₂
MW: 188.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16787128
CAS Number: 170432-54-9