2,2'-Difluorodiphenyldisulphide 98%
Supplier: Apollo Scientific
Warning
PC903987-100G PC903987-25G PC903987-5G
APOSPC903987-100GEA 720 EUR
APOSPC903987-100G APOSPC903987-25G APOSPC903987-5G
2,2'-Difluorodiphenyldisulphide 98%
2,2'-Difluorodiphenyldisulphide
Formula: C₁₂H₈F₂S₂
MW: 254.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029446
CAS Number: 14135-38-7