2-Fluoro-3-methyl-6-nitroaniline 95%
Supplier: Apollo Scientific
PC905356-1G PC905356-5G
APOSPC905356-1GEA 322 EUR
APOSPC905356-1G APOSPC905356-5G
2-Fluoro-3-methyl-6-nitroaniline 95%
2-Fluoro-3-methyl-6-nitroaniline
Formula: C₇H₇FN₂O₂
MW: 170.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18397069
CAS Number: 1261676-68-9