N-{[3-(Trifluoromethyl)pyridin-2-yl]methylidene}hydroxylamine 96%
Supplier: Apollo Scientific
PC910761-1G PC910761-5G
APOSPC910761-1GEA 219 EUR
APOSPC910761-1G APOSPC910761-5G
N-{[3-(Trifluoromethyl)pyridin-2-yl]methylidene}hydroxylamine 96%
N-{[3-(Trifluoromethyl)pyridin-2-yl]methylidene}hydroxylamine
Formula: C₇H₅F₃N₂O
MW: 190.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20277973
CAS Number: 1280538-35-3